Assay Details
| Gene Symbol: C25E10.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383365.1 | 3 | 364 | 70 | ||
| Gene Symbol: swm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072945.3 | NP_505346.1 | 4 | 444 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383365.1 transcripts.
Set Membership: |
|

C25E10.8
Genomic Map