Assay Details
| Gene Symbol: snf-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061509.9 | NP_493910.2 | 1 - 2 | 99 | 77 | |
| GenBank mRNA | DQ118731.1 | - | 1 - 2 | 141 | 77 | |
| DQ482733.1 | - | 1 - 2 | 121 | 77 | ||
More Information
Set Membership: |
|

snf-3
Genomic Map