Assay Details
| Gene Symbol: eri-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318309.3 | NP_001305238.1 | 1 - 2 | 313 | 148 | |
| NM_061517.8 | NP_493918.2 | 5 - 6 | 873 | 148 | ||
More Information
Set Membership: |
|

eri-3
Genomic Map