Assay Details
| Gene Symbol: fcho-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061546.4 | NP_493947.1 | 6 - 7 | 966 | 101 | |
| NM_061547.5 | NP_493948.1 | 5 - 6 | 902 | 101 | ||
More Information
Set Membership: |
|

fcho-1
Genomic Map