Assay Details
| Gene Symbol: fcho-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061546.4 | NP_493947.1 | 8 - 9 | 1739 | 76 | |
| NM_061547.5 | NP_493948.1 | 7 - 8 | 1675 | 76 | ||
More Information
Set Membership: |
|

fcho-1
Genomic Map