Assay Details
| Gene Symbol: aat-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061559.4 | NP_493960.3 | 7 - 8 | 1063 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aat-7| Gene Symbol: aat-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061559.4 | NP_493960.3 | 7 - 8 | 1063 | 61 | |
Set Membership: |
|