Assay Details
| Gene Symbol: nol-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377698.1 | NP_001364780.1 | 3 - 4 | 608 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nol-6| Gene Symbol: nol-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377698.1 | NP_001364780.1 | 3 - 4 | 608 | 93 | |
Set Membership: |
|