Assay Details
| Gene Symbol: fbxb-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061640.4 | NP_494041.2 | 3 - 4 | 1035 | 142 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fbxb-18| Gene Symbol: fbxb-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061640.4 | NP_494041.2 | 3 - 4 | 1035 | 142 | |
Set Membership: |
|