Assay Details
| Gene Symbol: gba-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047285.3 | NP_001040750.1 | 9 - 10 | 1281 | 118 | |
| NM_001047286.3 | NP_001040751.1 | 7 - 8 | 916 | 118 | ||
More Information
Set Membership: |
|

gba-1
Genomic Map