Assay Details
| Gene Symbol: cbs-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061814.4 | NP_494215.2 | 3 - 4 | 1017 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cbs-2| Gene Symbol: cbs-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061814.4 | NP_494215.2 | 3 - 4 | 1017 | 95 | |
Set Membership: |
|