Assay Details
| Gene Symbol: sri-77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061903.1 | NP_494304.1 | 3 - 4 | 507 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-77| Gene Symbol: sri-77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061903.1 | NP_494304.1 | 3 - 4 | 507 | 80 | |
Set Membership: |
|