Assay Details
| Gene Symbol: ptc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129178.3 | NP_001122650.1 | 12 - 13 | 4020 | 75 | |
| NM_061982.7 | NP_494383.2 | 13 - 14 | 4128 | 75 | ||
| NM_061983.8 | NP_494384.2 | 13 - 14 | 4108 | 75 | ||
More Information
Set Membership: |
|

ptc-3
Genomic Map