Assay Details
| Gene Symbol: ptc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129178.3 | NP_001122650.1 | 7 - 8 | 1166 | 105 | |
| NM_061982.7 | NP_494383.2 | 8 - 9 | 1265 | 105 | ||
| NM_061983.8 | NP_494384.2 | 8 - 9 | 1254 | 105 | ||
More Information
Set Membership: |
|

ptc-3
Genomic Map