Assay Details
| Gene Symbol: ptc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129178.3 | NP_001122650.1 | 9 - 10 | 2595 | 66 | |
| NM_061982.7 | NP_494383.2 | 10 - 11 | 2703 | 66 | ||
| NM_061983.8 | NP_494384.2 | 10 - 11 | 2683 | 66 | ||
More Information
Set Membership: |
|

ptc-3
Genomic Map