Assay Details
| Gene Symbol: irld-63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377738.1 | NP_001364806.1 | 2 - 3 | 347 | 130 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

irld-63| Gene Symbol: irld-63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377738.1 | NP_001364806.1 | 2 - 3 | 347 | 130 | |
Set Membership: |
|