Assay Details
| Gene Symbol: sri-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062030.1 | NP_494431.1 | 4 - 5 | 838 | 88 | |
| Gene Symbol: sri-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062031.5 | NP_494432.1 | 3 - 4 | 676 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_062031.5 transcripts.
Set Membership: |
|

sri-29
Genomic Map