Assay Details
| Gene Symbol: sri-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062032.5 | NP_494433.1 | 4 - 5 | 746 | 100 | |
| Gene Symbol: sri-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062033.2 | NP_494434.1 | 3 - 4 | 759 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_062033.2 transcripts.
Set Membership: |
|

sri-28
Genomic Map