Assay Details
| Gene Symbol: sri-42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062056.1 | NP_494457.1 | 1 - 2 | 146 | 178 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-42| Gene Symbol: sri-42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062056.1 | NP_494457.1 | 1 - 2 | 146 | 178 | |
Set Membership: |
|