Assay Details
| Gene Symbol: F18A12.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377755.1 | NP_001364708.1 | 4 - 5 | 649 | 161 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F18A12.7| Gene Symbol: F18A12.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377755.1 | NP_001364708.1 | 4 - 5 | 649 | 161 | |
Set Membership: |
|