Assay Details
| Gene Symbol: skpo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062139.8 | NP_494540.1 | 8 - 9 | 1207 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

skpo-2| Gene Symbol: skpo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062139.8 | NP_494540.1 | 8 - 9 | 1207 | 62 | |
Set Membership: |
|