Assay Details
| Gene Symbol: F54D10.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062262.3 | NP_494663.2 | 4 - 5 | 622 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F54D10.8| Gene Symbol: F54D10.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062262.3 | NP_494663.2 | 4 - 5 | 622 | 120 | |
Set Membership: |
|