Assay Details
| Gene Symbol: C17A2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062269.4 | NP_494670.1 | 4 - 5 | 572 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C17A2.2| Gene Symbol: C17A2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062269.4 | NP_494670.1 | 4 - 5 | 572 | 128 | |
Set Membership: |
|