Assay Details
| Gene Symbol: rpn-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062311.8 | NP_494712.1 | 3 - 4 | 381 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpn-11| Gene Symbol: rpn-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062311.8 | NP_494712.1 | 3 - 4 | 381 | 69 | |
Set Membership: |
|