Assay Details
| Gene Symbol: cct-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071121.7 | NP_503522.1 | 1 - 2 | 103 | 75 | |
| NM_182379.5 | NP_872179.1 | 1 - 2 | 97 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cct-7| Gene Symbol: cct-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071121.7 | NP_503522.1 | 1 - 2 | 103 | 75 | |
| NM_182379.5 | NP_872179.1 | 1 - 2 | 97 | 75 | ||
Set Membership: |
|