Assay Details
| Gene Symbol: nra-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070966.4 | NP_503367.1 | 3 - 4 | 665 | 67 | |
| NM_182391.2 | NP_872191.1 | 3 - 4 | 673 | 67 | ||
| Gene Symbol: flp-34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383273.1 | 3 - 4 | 664 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383273.1 transcripts.
Set Membership: |
|

nra-4
Genomic Map