Assay Details
| Gene Symbol: cpna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062336.5 | NP_494737.1 | 6 - 7 | 2266 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpna-1| Gene Symbol: cpna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062336.5 | NP_494737.1 | 6 - 7 | 2266 | 127 | |
Set Membership: |
|