Assay Details
| Gene Symbol: C01F1.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062352.8 | NP_494753.3 | 2 - 3 | 581 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C01F1.6| Gene Symbol: C01F1.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062352.8 | NP_494753.3 | 2 - 3 | 581 | 75 | |
Set Membership: |
|