Assay Details
| Gene Symbol: mlt-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062376.6 | NP_494777.1 | 4 - 5 | 766 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlt-7| Gene Symbol: mlt-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062376.6 | NP_494777.1 | 4 - 5 | 766 | 73 | |
Set Membership: |
|