Assay Details
| Gene Symbol: bpnt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062379.6 | NP_494780.1 | 1 - 2 | 134 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bpnt-1| Gene Symbol: bpnt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062379.6 | NP_494780.1 | 1 - 2 | 134 | 100 | |
Set Membership: |
|