Assay Details
| Gene Symbol: fut-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062422.8 | NP_494823.2 | 3 - 4 | 363 | 90 | |
| GenBank mRNA | AJ745074.1 | - | 3 - 4 | 361 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fut-6| Gene Symbol: fut-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062422.8 | NP_494823.2 | 3 - 4 | 363 | 90 | |
| GenBank mRNA | AJ745074.1 | - | 3 - 4 | 361 | 90 | |
Set Membership: |
|