Assay Details
| Gene Symbol: fut-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062422.8 | NP_494823.2 | 6 - 7 | 681 | 119 | |
| GenBank mRNA | AJ745074.1 | - | 6 - 7 | 679 | 119 | |
More Information
Set Membership: |
|

fut-6
Genomic Map