Assay Details
| Gene Symbol: fut-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062422.8 | NP_494823.2 | 9 - 10 | 1031 | 141 | |
| GenBank mRNA | AJ745074.1 | - | 9 - 10 | 1029 | 141 | |
More Information
Set Membership: |
|

fut-6
Genomic Map