Assay Details
| Gene Symbol: shc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383823.1 | 11 - 12 | 1827 | 68 | ||
| NM_062514.4 | NP_494915.2 | 11 - 12 | 1999 | 68 | ||
| Gene Symbol: T27F7.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383825.1 | 3 - 4 | 627 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383825.1 transcripts.
Set Membership: |
|

shc-2
Genomic Map