Assay Details
| Gene Symbol: shc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383823.1 | 4 - 5 | 696 | 74 | ||
| NM_062514.4 | NP_494915.2 | 4 - 5 | 868 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

shc-2| Gene Symbol: shc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383823.1 | 4 - 5 | 696 | 74 | ||
| NM_062514.4 | NP_494915.2 | 4 - 5 | 868 | 74 | ||
Set Membership: |
|