Assay Details
| Gene Symbol: shc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383823.1 | 6 - 7 | 934 | 78 | ||
| NM_001383824.1 | 6 - 7 | 920 | 78 | |||
| NM_062514.4 | NP_494915.2 | 6 - 7 | 1106 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383824.1 transcripts.
Set Membership: |
|

shc-2
Genomic Map