Assay Details
| Gene Symbol: wdr-83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062525.5 | NP_494926.1 | 3 - 4 | 534 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdr-83| Gene Symbol: wdr-83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062525.5 | NP_494926.1 | 3 - 4 | 534 | 125 | |
Set Membership: |
|