Assay Details
| Gene Symbol: dsh-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062536.9 | NP_494937.4 | 7 - 8 | 1857 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dsh-2| Gene Symbol: dsh-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062536.9 | NP_494937.4 | 7 - 8 | 1857 | 91 | |
Set Membership: |
|