Assay Details
| Gene Symbol: pld-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062538.7 | NP_494939.1 | 1 - 2 | 142 | 76 | |
| GenBank mRNA | AB028889.1 | - | 1 - 2 | 164 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pld-1| Gene Symbol: pld-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062538.7 | NP_494939.1 | 1 - 2 | 142 | 76 | |
| GenBank mRNA | AB028889.1 | - | 1 - 2 | 164 | 76 | |
Set Membership: |
|