Assay Details
| Gene Symbol: pld-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062538.7 | NP_494939.1 | 2 - 3 | 308 | 119 | |
| GenBank mRNA | AB028889.1 | - | 2 - 3 | 330 | 119 | |
More Information
Set Membership: |
|

pld-1
Genomic Map