Assay Details
| Gene Symbol: dsh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383828.1 | 2 - 3 | 240 | 84 | ||
| NM_001383829.1 | 1 - 2 | 140 | 84 | |||
| NM_062577.8 | NP_494978.4 | 2 - 3 | 236 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383829.1 transcripts.
Set Membership: |
|

dsh-1
Genomic Map