Assay Details
| Gene Symbol: dsh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383828.1 | 4 - 5 | 482 | 75 | ||
| NM_001383829.1 | 3 - 4 | 382 | 75 | |||
| NM_062577.8 | NP_494978.4 | 4 - 5 | 478 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383829.1 transcripts.
Set Membership: |
|

dsh-1
Genomic Map