Assay Details
| Gene Symbol: dsh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313591.3 | NP_001300520.1 | 2 - 3 | 166 | 79 | |
| NM_001313592.3 | NP_001300521.1 | 2 - 3 | 1591 | 79 | ||
| NM_001313593.3 | NP_001300522.1 | 2 - 3 | 166 | 79 | ||
| NM_001383828.1 | 7 - 8 | 869 | 79 | |||
| NM_001383829.1 | 7 - 8 | 2324 | 79 | |||
| NM_062577.8 | NP_494978.4 | 7 - 8 | 865 | 79 | ||
More Information
Set Membership: |
|

dsh-1
Genomic Map