Assay Details
| Gene Symbol: dsh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313591.3 | NP_001300520.1 | 3 - 4 | 295 | 100 | |
| NM_001313592.3 | NP_001300521.1 | 3 - 4 | 1720 | 100 | ||
| NM_001313593.3 | NP_001300522.1 | 3 - 4 | 295 | 100 | ||
| NM_001383828.1 | 8 - 9 | 998 | 100 | |||
| NM_001383829.1 | 8 - 9 | 2453 | 100 | |||
| NM_062577.8 | NP_494978.4 | 8 - 9 | 994 | 100 | ||
More Information
Set Membership: |
|

dsh-1
Genomic Map