Assay Details
| Gene Symbol: dsh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313591.3 | NP_001300520.1 | 4 - 5 | 534 | 117 | |
| NM_001313592.3 | NP_001300521.1 | 4 - 5 | 1959 | 117 | ||
| NM_001313593.3 | NP_001300522.1 | 4 - 5 | 534 | 117 | ||
| NM_001383828.1 | 9 - 10 | 1237 | 117 | |||
| NM_001383829.1 | 9 - 10 | 2692 | 117 | |||
| NM_062577.8 | NP_494978.4 | 9 - 10 | 1233 | 117 | ||
More Information
Set Membership: |
|

dsh-1
Genomic Map