Assay Details
| Gene Symbol: ptr-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062585.6 | NP_494986.2 | 10 - 11 | 2282 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-6| Gene Symbol: ptr-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062585.6 | NP_494986.2 | 10 - 11 | 2282 | 101 | |
Set Membership: |
|