Assay Details
| Gene Symbol: mog-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062618.7 | NP_495019.1 | 2 - 3 | 431 | 136 | |
| GenBank mRNA | AF286899.1 | - | 2 - 3 | 438 | 136 | |
More Information
Set Membership: |
|

mog-5
Genomic Map