Assay Details
Gene Symbol: dct-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062630.5 | NP_495031.2 | 3 - 4 | 351 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
