Assay Details
| Gene Symbol: nmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062632.7 | NP_495033.2 | 15 - 16 | 2403 | 119 | |
| GenBank mRNA | AF318613.2 | - | 15 - 16 | 2401 | 119 | |
More Information
Set Membership: |
|

nmr-1
Genomic Map