Assay Details
| Gene Symbol: nmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062632.7 | NP_495033.2 | 2 - 3 | 216 | 77 | |
| GenBank mRNA | AF318613.2 | - | 2 - 3 | 214 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nmr-1| Gene Symbol: nmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062632.7 | NP_495033.2 | 2 - 3 | 216 | 77 | |
| GenBank mRNA | AF318613.2 | - | 2 - 3 | 214 | 77 | |
Set Membership: |
|