Assay Details
| Gene Symbol: sam-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381430.2 | NP_001367525.1 | 3 - 4 | 574 | 68 | |
| Gene Symbol: F59E12.17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383848.1 | 3 - 4 | 574 | 68 | ||
| Gene Symbol: ddl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383847.1 | 3 - 4 | 578 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383847.1 transcripts.
Set Membership: |
|

sam-4
Genomic Map